Learn More
Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Description
It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 98.0% min. |
Solubility Information | Soluble in water. Slightly soluble in alcohol. |
Formula Weight | 174.18 |
Grade | ACS |
Sensitivity | Hygroscopic |
Appearance | White powder |
Chemical Name or Material | Potassium hydrogen phosphate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S18K027 | Certificate of Analysis | 30/09/2024 | 011593.A1,011593.30,011593.A7 |
U21K048 | Certificate of Analysis | 24/09/2024 | 011593.A1,011593.30,011593.A7 |
10250996 | Certificate of Analysis | 30/08/2024 | 011593.A1,011593.30,011593.A7 |
10251481 | Certificate of Analysis | 30/08/2024 | 011593.A1,011593.30,011593.A7 |
10249110 | Certificate of Analysis | 19/01/2024 | 011593.A1,011593.30,011593.A7 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.