missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydrogen phthalate, 99+%, Thermo Scientific Chemicals Product Code.: 10516412

Potassium hydrogen phthalate, 99+%, Thermo Scientific Chemicals

Product Code. 10516412
500 g, Glass bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10516412

Brand: Thermo Scientific Acros 417955000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasicShow MoreShow Less
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 295.0°C to 300.0°C
Color Red
pH 4 to 4.02 (0.05M soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula 2-(HO2C)C6H4CO2K
Quantity 500 g
Beilstein 09, 791
Merck Index 15, 7733
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 330g/L water (100°C), slightly soluble in alcohol
Formula Weight 204.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Potassium hydrogen phthalate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468118 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number417950025,417955000
Lot NumberA0452323 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number417950025,417955000
Lot NumberA0450649 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number417950025,417955000
Lot NumberA0375708 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number417950025,417955000
Lot NumberA0445192 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number417950025,417955000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydrogen phthalate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.