missing translation for 'onlineSavingsMsg'
Learn More
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals Product Code.: 10713504

Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals

Product Code. 10713504
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10713504

Brand: Thermo Scientific Acros 168880250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buokShow MoreShow Less
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]

Specifications

Melting Point 240.0°C to 258.0°C
Color White
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula (CH3)3COK
Quantity 25 g
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene
Formula Weight 112.21
Percent Purity 98+%
Grade Pure
Physical Form Powder
Chemical Name or Material Potassium tert-butoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0470645 Certificate TypeCertificate of Analysis Date11/04/2025 Catalog Number168880250,168881000,168880025,168885000,168880050
Lot NumberA0463889 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168880250,168881000,168880025,168885000,168880050
Lot NumberA0463889 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number168880250,168881000,168880025,168885000,168880050
Lot NumberA0462837 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number168880250,168881000,168880025,168885000,168880050
Lot NumberA0458873 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number168880250,168881000,168880025,168885000,168880050
compliance-icons compliance-icons
Product Identifier
  • Potassium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.