Learn More
Thermo Scientific™ Potato Dextrose Agar
Isolate and enumerate yeasts and molds from dairy and foodstuff with this ready-to-use medium.
Brand: Thermo Scientific™ PO0186A
12.57 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolate and enumerate yeasts and molds from dairy products1, the surface of fresh meats, cured meats and sausage products and other foods2 with Thermo Scientific™ Oxoid™ Potato Dextrose Agar. This medium is suitable for the detection and enumeration of heat resistant molds in thermally processed fruits and fruit products2. The agar used in Potato Dextrose Agar is carefully screened to ensure correct pigment production by fungi such as Fusaria spp.
Use Potato Dextrose Agar for detection and enumeration of yeasts and molds in foods.
- Suitable for detection of heat resistant molds in thermally processed fruits and fruit products.
- Suitable medium for the preparation of Aspergillus niger
- Conforms to EP/USP/JP Microbial Limit Test standards3,4,5.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Light Straw | |
Potato Dextrose Agar | |
United Kingdom | |
19g±2.0.g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
10 x 90mm plates, film wrapped |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4498687 | Certificate of Analysis | 19/03/2025 | PO0186A |
4498688 | Certificate of Analysis | 19/03/2025 | PO0186A |
4497407 | Certificate of Analysis | 17/03/2025 | PO0186A |
4497212 | Certificate of Analysis | 11/03/2025 | PO0186A |
4495855 | Certificate of Analysis | 09/03/2025 | PO0186A |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
10Minutes
35Seconds
8
Your input is important to us. Please complete this form to provide feedback related to the content on this product.