missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Power-Bind™ Streptavidin Coated Particles

Bind a variety of biotinylated ligands, including DNA/RNA, oligos, antibodies and proteins with Thermo Scientific™ Power-Bind Streptavidin Coated Particles.

Brand:  Thermo Scientific™ 29001801011150

 View more versions of this product

Product Code. 11844992

  • £101.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Includes Certificate of Analysis
Power-Bind Streptavidin Coated Particles
Biotinylated Ligand Binding, Diagnostic Assays
Streptavidin-Coated Particles
0.05% Na Az
Power-Bind evaluation samples are packaged in 1mL bottles at 1% solids (10 mg/mL). Select individual lots to evaluate.
Guanidinium thiocyanate; DMF and DMSO to 100% Sonication; environments PCR temperature cycling
2°C to 8°C
White or Colorless
Polystyrene
Package Insert and Certificate of Analysis
1 mL
Power-Bind Streptavidin Coated particles are available in different concentrations and package sizes upon request.
0.85 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number257317 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number29001801011150
Lot Number267529 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number29001801011150
Lot Number245914 Certificate TypeCertificate of Analysis Date13/01/2022 Catalog Number29001801011150
Lot Number238542 Certificate TypeCertificate of Analysis Date06/04/2021 Catalog Number29001801011150
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Power-Bind™ Streptavidin Coated Particles > Dia: 0.85μm; 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.