missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Power Blotter Select Transfer Stacks, PVDF, mini

Transfer proteins rapidly and efficiently with these ready-to-use high performance transfer stacks for Invitrogen Power Blotter.

Brand:  Invitrogen™ PB5240

307.80 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15876162

  • £380.00 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.


Polyvinylidene diflouride (PVDF) membrane:

  • 0.2 μm pore size, low fluorescence
  • Pre-activated and ready for use without any alcohol pretreatment.  Compatible with
  • commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling.
  • Poteins bind as hydrophobic interactions
  • Protein-binding capacity is 240 μg/cm2

Rapid, efficient and even transfer:

  • Ready-to-use transfer stack takes little set-up time
  • Protein transfers in as little as 7 minutes
  • Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity

Increased reproducibility:

  • Preassembled stacks provide consistent performance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 cm
Transfer Stack
0.2 μm
Room Temperature
Power Blotter
8 cm
PVDF
40 mini-size transfer stacks (8 cm x 8 cm)
8 cm x 8 cm
40 Blots
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.