Learn More
Invitrogen™ Power Blotter Select Transfer Stacks, PVDF, mini
Transfer proteins rapidly and efficiently with these ready-to-use high performance transfer stacks for Invitrogen Power Blotter.
£116.00 - £400.00
Specifications
Length (Metric) | 8 cm |
---|---|
Width (Metric) | 8 cm |
Format | Transfer Stack |
Material | PVDF |
Pore Size | 0.2 μm |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15866162
|
Invitrogen™
PB5210 |
10 Blots |
£116.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15876162
|
Invitrogen™
PB5240 |
40 Blots |
£400.00
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.
Polyvinylidene diflouride (PVDF) membrane:
- 0.2 μm pore size, low fluorescence
- Pre-activated and ready for use without any alcohol pretreatment. Compatible with
- commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling.
- Poteins bind as hydrophobic interactions
- Protein-binding capacity is 240 μg/cm2
Rapid, efficient and even transfer:
- Ready-to-use transfer stack takes little set-up time
- Protein transfers in as little as 7 minutes
- Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity
Increased reproducibility:
- Preassembled stacks provide consistent performance
Specifications
8 cm | |
Transfer Stack | |
0.2 μm | |
8 cm x 8 cm |
8 cm | |
PVDF | |
Room Temperature | |
Power Blotter |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.