Learn More
Fisherbrand™ Power Supply, Mini 300V Plus
Provides up to 300V (volts) and 400mA of power. Fisherbrand™ Power Supply, 300V Mini Plus includes dual voltage compatibility.
Brand:  Fisherbrand™ MINI300VPLUS
Product Code. 12643546
  • £570.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Enhanced built-in safety features
  • Three digit LED display
  • Alarm function
  • Wipe clean polycarbonate housing