missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Beatus Bottle Top Dispenser

Robust and reliable dispensing; Fisherbrand™ Beatus Bottle Top Dispenser has re-circulation valve for re-directing the liquid into the bottle for bubble free dispensing without any loss of reagent.

Brand:  Fisherbrand™ BEAT2.5

 View more versions of this product

Product Code. 15518901

  • ÂŁ481.00 / Each
Estimated Shipment: 17-06-2025
Explore more special offers
Add to basket

Description

Description

  • With re-circulation valve
  • Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
  • Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
  • Specially designed adaptor allows 360° rotation providing full flexibility
  • Springless, PTFE valve manifold ensures high chemical compatibility and jamming free valve function
  • PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
  • Adaptors to fit most of the laboratory reagent bottles
  • Calibration tool provided for convenient and quick in-lab user re-calibration

Specifications:

  • Piston: PTFE and ETP
  • Valve housing: PFA
  • Re-Circulation valve housing: PFA
  • Valve assembly: Borosilicate glass and seat
  • Discharge assembly: PTFE
  • Delivery and Inlet tube: FEP
  • Compatibility: With all reagents except HF
Specifications

Specifications

0.6% (+/-0.015)
Autoclavable (121°C, 15psi for 10-15 min.)
Borosilicate Glass
0.05 mL
Bottletop Dispenser
ISO 8655 standards
0.25 to 2.5 mL
BEAT-2.5
Product Suggestions

Product Suggestions

Videos

Videos

  • Talking Points - Fisherbrand Bottle Top Dispensers
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Talking Points - Fisherbrand Bottle Top Dispensers

    SDS
    Documents Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Fisherbrand™ Beatus Bottle Top Dispenser > Increment: 0.05mL; Volume: 0.25 - 2.5mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.