missing translation for 'onlineSavingsMsg'
Learn More
Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11351398

Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11351398
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11351398

Brand: Thermo Scientific Alfa Aesar 010305.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Gray
Quantity 25 x 25 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.27g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Praseodymium foil, 0.25mm (0.01 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU02K015 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number010305.FF
Lot NumberB09X051 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010305.FF
Lot NumberL26L27 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010305.FF
Lot NumberU27E042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010305.FF
Lot NumberL14P51 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010305.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.