Learn More
JULABO™ PRESTO™ W80 Temperature Control System/Process System
Made for highly precise temperature control and rapid temperature changes. Allows extremely fast compensation of exothermic and endothermic reactions. Lab users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure.
Brand:  JULABO™ 9421801
Product Code. 15274363
  • £39150.00 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilization in your lab
  • 5,7” industrial color TFT touch screen
  • USB (Host und Device)
  • Ethernet
  • SD-Card slot
  • RS232
  • Modbus
  • RS485 (Accessory)
  • Profibus DP (Accessory)
  • Stand-by input
  • Water cooled
  • Permanent internal monitoring and self-lubricating pumps contribute to a long service life
Specifications
Specifications
Classification III (FL) according to DIN 12876-1
250°C
65 x 43 x 126cm
159 kg
Temperature Control System
230 V
RS232, SD memory card, USB, Ethernet, Modbus
-80°C to +250°C
PRESTO™ W80
TFT Touchscreen
Ideal solution for highly precise, external temperature application
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
JULABO™ PRESTO™ W80 Temperature Control System/Process System > 2-stage water Cooling; Range: -80 to 250°C; Heating Capacity: 1.8kW