missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Supplied with 70mm x 70mm and 70mm x 100mm gel trays
- Economic low gel and buffer volumes
- Small footprint
Specifications
32 Samples (max., for 70mm x 70mm tray); 64 Samples (max., for 70mm x 100mm tray) | |
70mm x 70mm and 70mm x 100mm gel trays | |
210 x 90 x 90 mm |
70 x 70 mm | |
225 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
-
Fisherbrand Meet the Family
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ MultiSUB Mini Horizontal Gel System > Length: 210mm
Spot an opportunity for improvement?Share a Content Correction