Learn More
JULABO™ PRESTO™ A30 Temperature Control System/Process System
Julabo™ PRESTO™ A30 Temperature Control System/Process System allows extremely fast compensation of exothermic and endothermic reactions. Lab users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure.
Brand:  JULABO™ 9420300
Product Code. 15224363
  • £16020.00 / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilization in your lab
  • 5,7” industrial color TFT touch screen
  • USB (Host und Device)
  • Ethernet
  • SD-Card slot
  • RS232
  • Modbus
  • RS485 (Accessory)
  • Profibus DP (Accessory)
  • Stand-by input
  • Removable venting grid for simplified removal of dust
  • Permanent internal monitoring and self-lubricating pumps contribute to long service life
Specifications
Specifications
Classification III (FL) according to DIN 12876-1
RS232, SD memory card, USB, Ethernet, Modbus
250°C
25 cm
PRESTO™ A30
TFT Touchscreen
Ideal solution for highly precise, external temperature application
62 cm
59 cm
-30°C to +250°C
59 x 25 x 62 cm
62 kg
Temperature Control System
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
JULABO™ PRESTO™ A30 Temperature Control System/Process System > 1-stage air Cooling; Range: -30 to 250°C; Heating Capacity: 2.7kW