missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ProBond™ Nickel-Chelating Resin
A nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence.
Brand: Invitrogen™ R80115
This item is not returnable.
View return policy
Description
- ProBond™ Nickel-Chelating Resin is a nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
- Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine
- One-step purification can be performed under both native and denaturing conditions
- ProBond™ Resin uses the chelating ligand iminodiacetic acid (IDA) coupled to a highly cross-linked 6% agarose resin
- Suitable for use in FPLC, batch, and gravity flow applications
- Guaranteed stable for 6 months when properly stored at 4°C

Specifications
The ProBond™ resin is pre-charged and able to bind 1-5 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 20% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. ProBond™ resin is guaranteed stable for 6 months when properly stored. | |
ProBond™ | |
150 mL | |
Affinity Column |
Nickel-Chelating | |
Resin | |
Suspension |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ ProBond™ Nickel-Chelating Resin > 150 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction