missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Procaine hydrochloride, 99% Product Code.: 10542421

Thermo Scientific Chemicals Procaine hydrochloride, 99%

Product Code. 10542421
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10542421

Brand: Thermo Scientific Chemicals 207311000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolorShow MoreShow Less
PubChem CID 5795
ChEBI CHEBI:8431
IUPAC Name 2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 154.0°C to 158.0°C
Color Colorless to White
pH 5 to 6.5 (2% w/v aq. soln.)
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NC6H4CO2CH2CH2N(C2H5)2·HCl
Quantity 100 g
Beilstein 14, 424
Merck Index 15, 7875
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform, practically insoluble in ether
Formula Weight 272.77
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Procaine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466253 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number207311000,207315000
Lot NumberA0464387 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number207311000,207315000
Lot NumberA0463797 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number207311000,207315000
Lot NumberA0461399 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number207311000,207315000
Lot NumberA0457716 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number207311000,207315000
compliance-icons
Product Identifier
  • Procaine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Procaine hydrochloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.