Learn More
Invitrogen™ ProcartaPlex Mouse Basic Kit
Ideal solution for assessing multiple protein biomarkers in a single sample. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates. Explore our broad portfolio of pre-configures panels and combinable single analytes.
Brand:  Invitrogen™ EPX010-20440-901
Product Code. 15502137
  • £210.00 / 96 tests
Explore more special offers
Includes:
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent Mouse
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • This kit does not contain a Multiscreen Filter Plate
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Basic and Setup Kits
Mouse
See Certificate of Analysis
ProcartaPlex™
96 Tests
Combinable
Luminex™ Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number438175-000 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number
Lot Number438175-000 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number
Lot Number438175-000 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberEPX010-20440-901
Lot Number421051-000 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberEPX010-20440-901
Lot Number409799-000 Certificate TypeCertificate of Analysis Date11/05/2024 Catalog NumberEPX010-20440-901
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ ProcartaPlex Mouse Basic Kit > ProcartaPlex Mouse Basic Kit