Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
Brand:  WTW™ 209716
Additional Details : Weight : 11.00000kg
 View more versions of this product
Product Code. 16391736
  • £6190.00 / Pack of 12
Estimated Shipment: 22-08-2024
to see stock.
Description
Description
  • OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
  • Measuring ranges up to 4000 mg/L BOD possible
  • No need to convert data - direct input of sample volume
  • Direct display of the BOD value on the measuring head screen
  • Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
Specifications
OxiTop™-IDS 12
Backlit Graphic LCD
Measuring ranges up to 4000 mg/L BOD possible
Twelve OxiTop™-IDS measuring heads