missing translation for 'onlineSavingsMsg'
Learn More
uvex Profastrong NF33 Chemical Protection Gloves Product Code.: 15386922
Add to basket

uvex Profastrong NF33 Chemical Protection Gloves

Product Code. 15386922 Shop All Uvex Products
8
Click to view available options
Size:
7
8
9
10

Product Code. 15386922

Brand: uvex 6012202

Add to basket
Add to basket

Sensitive NBR chemical protection gloves feature a flocked cotton lining for comfort and provide a high degree of mechanical abrasion resistance.

  • Sensitive NBR chemical protection gloves with flocked cotton lining for comfort
  • Embossed structure on the palm enhances grip capabilities in damp and wet areas
  • Nitrile rubber is resistant to oils, grease, acids, and alkalis
  • Gloves provide a tactile feel, secure fit, and mechanical abrasion resistance
  • Ideal for use in various industries including automotive, chemical, and printing
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Latex Free Yes
Material NBR
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 4101X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLOT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Profastrong NF33
Length (Metric) 33 cm
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex Profastrong NF33 Chemical Protection Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.