Learn More
Sartorius PROLINE™ Plus Fixed Volume, Single-Channel Mechanical Pipetters
GREENER_CHOICEBIOPHARMA
Robust yet lightweight and easy to use
£188.00 - £249.00
Specifications
Certifications/Compliance CE
Autoclavable Autoclavable
Disposable Reusable
Material Polypropylene
Number of Channels 1
View More Specs
Products 13
Product Code Brand Accuracy Color Volume (Metric) Model Price Quantity & Availability  
Product Code Brand Accuracy Color Volume (Metric) Model Price Quantity & Availability  
10774112
GREENER_CHOICEBIOPHARMA
View Documents
Sartorius
728530
0.6% Yellow 20 μL Fix 20 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10784112
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728535
0.5% Yellow 25 μL Fix 25 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10763542
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728550
0.5% Yellow 100 μL Fix 100 μL
£249.00
Each
In Stock
to see stock.
Add to basket
10047859
BIOPHARMA
View Documents Promotion Details
Sartorius
728545
0.5% Yellow 50 μL Fix 50 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10038149
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728580
0.3% Green 5000 μL F5
£194.00
Each
In Stock
to see stock.
Add to basket
10322310
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728590
0.6% Red 10 mL F10
£194.00
Each
In Stock
to see stock.
Add to basket
10199599
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728520
0.8% Gray 10 μL Fix 10 μL
£249.00
Each
In Stock
to see stock.
Add to basket
10577600
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728570
0.3% Blue 1000 μL Fix 1000 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10312310
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728575
0.3% Green 2000 μL F2
£194.00
Each
In Stock
to see stock.
Add to basket
10047669
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728567
0.3% Blue 500 μL Fix 500 μL
£194.00
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10322500
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
728560
0.4% Yellow 200 μL Fix 200 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10821701
BIOPHARMA
View Documents Promotion Details
Sartorius
728565
0.4% Blue 250 μL Fix 250 μL
£194.00
Each
In Stock
to see stock.
Add to basket
10058239
BIOPHARMA
View Documents Promotion Details
Sartorius
728515
1.3% Gray 5 μL Fix 5 μL
£188.00
Each
In Stock
to see stock.
Add to basket
Description
Description
High quality and light pipetting forces provide a pleasant pipetting experience. Covering the full range of 0.1μl to 10mL, the Proline Plus is suitable for a wide range of everyday laboratory applications.
  • Fully autoclavable
  • High chemical and UV resistance
  • Volume setting by click stops
  • Suitable for both left and right-handed operation
  • Compatible with a wide range of tips
  • Compatible with most commonly used lab tubes due to slim tip ejector collar
  • Safe-Cone Filter prevents contamination and damage of pipetter
  • Volume range color-coding for easy pipetter and tip selection
  • Easy cleaning: only 2-3 parts to disassemble
Certifications
CE marked
Warranty
2 years
Specifications
Specifications
CE
Reusable
1
Ergonomic
Yes
Autoclavable
Polypropylene
Non-sterile
Slim tip ejector collar, safe cone filter
2 Years
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius PROLINE™ Plus Fixed Volume, Single-Channel Mechanical Pipetters