Learn More
Invitrogen™ ProLong™ Diamond Antifade Mountant with SYTOX™ Deep Red
ProLong Diamond Antifade Mountant is compatible with almost any fluorescent dye or fluorescent protein across visible and IR spectrums. This mountant contains SYTOX Deep Red DNA stain for staining the nuclei of fixed cells.
£66.75 - £207.00
Specifications
Product Type | Antifade Mountant |
---|---|
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
Reagent Type | Mounting Solution, Antifade Solution |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17108302
|
Invitrogen™
P36990 |
ProLong™ Diamond Antifade Mountant with SYTOX™ Deep Red, 1 x 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17137388
|
Invitrogen™
P36990-5X2ML |
ProLong™ Diamond Antifade Mountant with SYTOX™ Deep Red, 5 x 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
ProLong Diamond mounting media is the ideal option for archiving samples with fluorescent dyes or fluorescent proteins. ProLong Diamond mountant cures at 1.47 RI and enables longer-term storage of your samples, while protecting them from photobleaching. ProLong Diamond Antifade Mountant comes ready to use in a 2 mL dropper bottle with SYTOX Deep Red DNA stain for nuclear counterstaining. ProLong Diamond Antifade Mountant with SYTOX Deep Red stain has an excitation of 660 nm when bound to DNA with an emission maximum at 682 nm detected using a Cy5/deep red traditional filter. Just apply a drop to the sample, add a coverslip, cure, and image. ProLong Diamond mountant does not discolor or shrink when cured and stored long term, making it possible to take high quality fluorescent images.
ProLong Diamond Antifade Mountant is compatible with a wide array of fluorescent dyes, including Alexa Fluor dyes, FITC, TRTC, Texas Red, Cy3, and Cy5. ProLong Diamond mounting media does not quench fluorophores, and its ability to provide brighter signal, against low background, helps ensure that you get the best-quality, high resolution images for publishing.
Specifications
Antifade Mountant | |
Approved for shipment at Room Temperature or on Wet Ice | |
Mounting Solution, Antifade Solution |