Learn More
Thermo Scientific™ ProPac™ Elite WCX HPLC Column
This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.
Brand: Thermo Scientific™ 302973
842.44 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.
The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.
Product Highlights- Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
- High efficiency with reproducible separa-tions
- High recovery with low carry-over
- Wide pH operating range: 2 – 12
- High temperature stability: up to 60°C
- High throughput
- Compatible with CX-1 pH Gradient Buffers

Specifications
Analytical Column | |
0.4 to 1.0 mL/min | |
4500 psi (310 bar) | |
5 μm | |
Nonporous | |
10°C to 60°C | |
ProPac | |
Ion Exchange | |
Bio-LC |
4 mm | |
50 mm | |
Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O | |
2 to 12 | |
1 Column | |
Carboxylate | |
Nonporous DVB Particles with Hydrophilic Coating | |
Ion Exchange | |
Responsibly packaged |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.