Learn More
Cytiva Protein G Sepharose™ 4 Fast Flow Media
Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.
Brand:  Cytiva 17-0618-01
 View more versions of this product
Product Code. 11524935
  • £656.00 / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2mg Protein G/mL Medium
90 μm
5 mL
Used in a range of diagnostic and research applications
4% Cross Linked Agarose
3 to 9
2°C to 8°C
Recombinant Protein G lacking albumin binding region