Learn More
Macherey-Nagel™ Protino Ni-TED Resin for His-tag Protein Purification
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications. The matrix of choice for highest protein purity as dry bulk material.
Brand: Macherey-Nagel™ 745200.30
Description
Protino™ NI-TED, the purer alternative for His-tag protein purification- Chelating group TED, one protein binding site
- High stability against reducing/chelating agents
- Low metal leaching
- High protein purity
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-TED resin
- Empty 96 well receiver plates available for use with Protino™ Ni-TED resin For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
10mg/g | |
0.5 to 1.0 mL/min./cm2 | |
Macroporous Silica | |
30 g Dry Silica Bulk Resin | |
Resin |
Batch | |
Buffers | |
145 psi | |
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications | |
TED (tris-carboxymethyl ethylene diamine) |
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- Respiratory sensitiser Category 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360-May damage fertility or the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : resin with Ni in nickel(II) complexes
Your input is important to us. Please complete this form to provide feedback related to the content on this product.