Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
£3.50 - £45.05
Products 12
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
11898882
View Documents
Fisherbrand™
001.808
1.5 mm 8 mm
£45.05
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10652401
View Documents
Fisherbrand™
001.810
3 mm 10 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11868882
View Documents
Fisherbrand™
001.803
3 mm 3 mm
£13.30
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11808892
View Documents
Fisherbrand™
001.813
3 mm 13 mm
£11.45
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11878882
View Documents
Fisherbrand™
001.805
2 mm 5 mm
£43.65
Pack of 10
In Stock
to see stock.
Add to basket
11828892
View Documents
Fisherbrand™
001.815
1.5 mm 15 mm
£20.60
Pack of 10
In Stock
to see stock.
Edge
Add to basket
12656316
View Documents
Fisherbrand™
001.820
3 mm 20 mm
£22.20
Pack of 10
In Stock
to see stock.
Edge
Add to basket
16245851
View Documents
Fisherbrand™
001.830
3 mm 30 mm
£20.30
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11888882
View Documents
Fisherbrand™
001.806
3 mm 6 mm
£26.40
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11858882
View Documents
Fisherbrand™
001.802
2 mm 2 mm
£28.15
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11818892
View Documents
Fisherbrand™
001.808.3
3 mm 8 mm
£32.21
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11888952
View Documents
Fisherbrand™
001.807
2 mm 7 mm
£17.45
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ PTFE Micro Magnetic Stir Bar