Learn More
Moldex™ Pura-Fit MoldexStations Product Code.: 12344759
Add to basket
Moldex™ Pura-Fit MoldexStations
Product Code. 12344759
Size: Large
Click to view available options
Size: Large
Product Code. 12344759
Brand: Moldex™ 772501
Add to basket
Add to basket
Plugs remain hygienic as the dispenser mechanism is automatically replaced with every refill
Made from recyclable material and with only 2% wastage the dispenser puts protection at the center of the workforce. Plugs remain hygienic as the dispenser mechanism is automatically replaced with every refill. Contains 250 pairs.
Specifications
Includes 250 Pairs of Moldex Pura-Fit earplugs
Size Large
Quantity 250/Pk.
Noise Reduction Rating (NRR) 36 dB
Product Type Pura-Fit MoldexStation
Product Content Correction
Product Title
Moldex™ Pura-Fit MoldexStations >