missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Purified Water for Microbiology

Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Purified Water for Microbiology.

£25.00 - £357.00

Specifications

Content And Storage 2 to 25°C
Manufacturing Origin United Kingdom
Product Type Purified Water
View More Specs

Products 7
Product Code Brand Fill Weight Price Quantity & Availability  
Product Code Brand Fill Weight Price Quantity & Availability  
12746919
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
BO0184B
3.0g + 0.3g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12736929
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
BO0209B
3.0g + 0.3g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
13275639
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
BO0209C
5.0g + 0.3g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12756929
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
BO0209M
100.0g + 1.5g
£80.15
Pack of 10
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Edge
Add to basket
 
12723586
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
EB0209B
3.0g + 0.3g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12947048
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
EB0209C
5.0g - 5.3g
£357.00
Pack of 200
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Add to basket
 
12766929
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
BO0209R
200.0g + 4.0g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Thermo Scientific™ Oxoid™ Purified Water. The bottles come with screw cap closure for ease of operation.

Use Purified Water as a diluting and rinsing fluid.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Specifications

2 to 25°C
United Kingdom
Purified Water
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 52 Weeks

SDS
Documents

Documents

Certificates