missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ pHPerch Benchtop Workstation

Keep buffers, wash bottles, and waste beakers organized and in easy reach with the pHPerch Benchtop Workstation.

Brand:  Cole-Parmer™ 63527-30

Product Code. 11716068

  • £267.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 1 year warranty.
Specifications

Specifications

Organizer
6 in.
12 in.
Buffers, Wash Bottles, Waste Beakers
10
33.02 cm
White
15.24 cm
30.48 cm
Small
13 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ pHPerch Benchtop Workstation > pHPerch Benchtop Workstation; Dimensions (D x W x H): 15.24 x 33.02 x 30.48 cm (6 x 13 x 12 in.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.