missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PVDF Transfer Membrane, 0.2μm, 26.5 cm x 3.75 m

PVDF Transfer Membrane, 0.2 μm, 26.5 cm x 3.75 m

Brand:  Thermo Scientific™ 88520

453.15 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.


Product Code. 10078559

  • £504.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for Western, Southern, and Northern blotting methods. This PVDF membrane has a 0.2-micron pore size and is ideal for colorimetric and chemiluminescent detection.

Features of PVDF membranes:

  • Western blotting—PVDF transfer membranes manufactured especially for protein transfer and Western blot applications
  • Compatible and durable—polyvinyl difluoride (PVDF) is compatible with most organic solvents, acids and mild bases; doesn't tear or become brittle like nitrocellulose
  • Specialized—use the 0.2μm variety for colorimetric and chemiluminescent detection

    Our PVDF (polyvinyl difluoride) membranes are more resistant to discoloration than other commercially available PVDF membranes and have better retention of adsorbed proteins than other types of transfer membranes used for Western blotting, including nitrocellulose. This membrane has lower background levels and increased sensitivity for fluorescent probing than regular PVDF or nitrocellulose membranes.

    Related Products
    PVDF Transfer Membrane, 0.45μm, 26.5 cm x 3.75 m
    PVDF Transfer Membrane, 0.45μm, 10 cm x 10 cm
    Low-Fluorescence PVDF Transfer Membrane, 0.2μm, 7 cm x 8.4 cm
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    PVDF Transfer Membrane, 0.2μm Pore Size, 26.5cm x 3.75m, Membranes are Made of High-Quality Polyvinylidene Difluoride and Provide High Binding Capacity for Proteins and Nucleic Acids for Western, Southern, and Northern Blotting Methods
    Protein assays and analysis, western blotting, protein biology
    1 Roll
    0.2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    • Multiple Manufacturers
      Advance your proteomics research with essential tools
      Promo Code: 25977

      Expires2025-05-31

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Thermo Scientific™ PVDF Transfer Membrane, 0.2μm, 26.5 cm x 3.75 m > 1 roll

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.