Learn More
Pyrex™ Borosilicate Glass Class A Certified Volumetric Flask
Manufactured from borosilicate glass for corrosion free-performance. Pyrex™ Borosilicate Glass Class A Certified Volumetric Flask features white enamel inscriptions for easy volume assessment. Includes polyethylene stopper. Conformity 'H' certified, complies with ISO 1042 and DIN 12664 and is calibrated to the tolerances required by the United States Pharmacopoeia (USP.)
£91.05 - £361.00
Specifications
Certifications/Compliance Class A, ISO 1042, DIN 12664, USP, Conformity 'H' certified
Closure Material Polyethylene
Description White enamel inscriptions
Material Borosilicate Glass
Graduated Yes
View More Specs
Products 14
Product Code Brand Capacity (Metric) Closure Size Height (Metric) Tolerance Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Height (Metric) Tolerance Price Quantity & Availability  
11353634
missing translation for 'viewDocuments'
Pyrex
3151/01WC
1 mL 7/16 65 mm ±0.02 mL
£158.00
Pack of 5
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
10059279
missing translation for 'viewDocuments'
Pyrex
3151/24WC
2000 mL 29/32 370 mm ±0.5 mL
£170.00
Each
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11363634
missing translation for 'viewDocuments'
Pyrex
3151/02WC
2 mL 7/16 65 mm ±0.02 mL
£197.00
Pack of 5
Estimated Shipment: 24-07-2025
to see stock.
Add to basket
10811601
missing translation for 'viewDocuments'
Pyrex
3151/26WC
5 L 34/35 475 mm ±1 mL
£218.40
Each
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11373634
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/04WC
5 mL 10/19 70 mm ±0.02 mL
£91.05
Pack of 5
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
11383634
missing translation for 'viewDocuments'
Pyrex
3151/06WC
10 mL 10/19 90 mm ±0.02 mL
£233.00
Pack of 5
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
11393634
missing translation for 'viewDocuments'
Pyrex
3151/08WC
20 mL 10/19 110 mm ±0.03 mL
£244.00
Pack of 5
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11303644
missing translation for 'viewDocuments'
Pyrex
3151/10WC
25 mL 10/19 110 mm ±0.03 mL
£99.00
Pack of 5
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11313644
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/12WC
50 mL 12/21 140 mm ±0.05 mL
£111.28
Pack of 5
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
11323644
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/14WC
100 mL 14/23 170 mm ±0.08 mL
£296.00
Pack of 5
In Stock
to see stock.
Edge
Add to basket
11333644
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/16WC
200 mL 14/23 210 mm ±0.1 mL
£361.00
Pack of 5
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11911528
missing translation for 'viewDocuments'
Pyrex
3151/18WC
250 mL 14/23 220 mm ±0.12 mL
£140.00
Pack of 2
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
11353644
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/20WC
500 mL 19/26 260 mm ±0.2 mL
£117.00
Pack of 2
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
11363644
missing translation for 'viewDocuments' Promotion Details
Pyrex
3151/22WC
1000 mL 24/29 300 mm ±0.3 mL
£142.00
Pack of 2
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
Description
Description
Manufactured from borosilicate glass for corrosion free-performance. Pyrex™ Borosilicate Glass Class A Certified Volumetric Flask features white enamel inscriptions for easy volume assessment. Includes polyethylene stopper. Conformity 'H' certified, complies with ISO 1042 and DIN 12664 and is calibrated to the tolerances required by the United States Pharmacopoeia (USP.)
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerances required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Each flask engraved with individual serial number and supplied with certificate of calibration traceable to National Standards
  • Conformity 'H' certified
  • Manufactured from borosilicate glass for corrosion-free performance
  • White enamel inscriptions
  • Polyethylene stopper
Specifications
Specifications
Class A, ISO 1042, DIN 12664, USP, Conformity 'H' certified
White enamel inscriptions
Yes
Polyethylene
Borosilicate Glass
A
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Class A Certified Volumetric Flask