Learn More
Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock. Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask complies with ISO 1773 standards. Excellent chemical durability for contamination-free performance. Includes white enamel marking spot while flat base prevents spillage.
£11.45 - £472.00
Specifications
Certifications/Compliance ISO 1773
Material Borosilicate Glass
Neck Style Narrow
Bottom Shape Flat
View More Specs
Products 11
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Neck Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Neck Height (Metric) Price Quantity & Availability  
10650191
View Documents
Pyrex
1070/38D
10 L 280 mm 57 mm 360 mm
£334.00
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
10009569
View Documents
Pyrex
1070/04D
100 mL 64 mm 18 mm 110 mm
£11.45
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
10211560
View Documents
Pyrex
1070/40D
12 L 300 mm 57 mm 465 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10662111
View Documents
Pyrex
1070/42D
20 L 341 mm 60 mm 558 mm
£472.00
Each
Estimated Shipment: 30-09-2025
to see stock.
Add to basket
10801601
View Documents
Pyrex
1070/28D
2000 mL 166 mm 45 mm 250 mm
£384.00
Pack of 10
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
12549085
View Documents
Pyrex
1070/02M
50 mL 50 mm 18 mm 100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10650001
View Documents
Pyrex
1070/36D
6 L 237 mm 57 mm 340 mm
£133.00
Each
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
12079002
View Documents
Pyrex
1070/12D
250 mL 85 mm 30 mm 140 mm
£144.00
Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12089002
View Documents
Pyrex
1070/18D
500 mL 105 mm 30 mm 170 mm
£147.00
Pack of 10
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
11982609
View Documents
Pyrex
1070/24D
1000 mL 131 mm 36 mm 200 mm
£217.00
Pack of 10
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
10640001
View Documents
Pyrex
1070/32D
4 L 207 mm 45 mm 300 mm
£135.00
Each
Estimated Shipment: 30-09-2025
to see stock.
Add to basket
Description
Description
  • Complies with ISO 1773
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination free performance
  • With white enamel marking spot
Specifications
Specifications
ISO 1773
Borosilicate Glass
Narrow
Flat