missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Thimble

Pyrex™ Borosilicate Glass Thimble is highly resistant to chemical attack.

Brand:  Pyrex 3840/02M

272.75 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15507730

  • £273.00 / Pack of 10
Estimated Shipment: 22-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Borosilicate glass with sintered glass disc to support solid sample
  • Useful for chemical applications where weighing extracted material is required
TRUSTED_SUSTAINABILITY
Specifications

Specifications

26 mm
Useful for chemical applications where it is required to weigh the extracted material
Sintered Glass disc
Filter Thimble
Borosilicate Glass
Porosity 1
95 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Borosilicate Glass Thimble > Disc Diameter: 20mm; Outside Diameter: 26mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.