Learn More
Corning™ PYREX™ Shorty Disposable Glass Serological Pipets, TD, Sterile, Plugged
Ideal for use in confined areas such as hoods
£118.96 - £886.45
Specifications
Aspirator Plug | Yes |
---|---|
Calibrated | Yes |
Material | Borosilicate Glass |
Graduated | Yes |
Tip Style | Round |
Product Code | Brand | Volume (Metric) | Graduations | Negative Graduations | Packaging Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Negative Graduations | Packaging Type | Price | Quantity & Availability | ||
10406653
|
Corning™
7078B-25 |
25 mL | 0.2 mL | 5 mL | Multi-pack |
£886.45
Pack of 400 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10496643
|
Corning™
7078B-10 |
10 mL | 0.2 mL | 4 mL | Multi-pack |
£659.12
Pack of 400 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10736323
|
Corning™
7078B-1 |
1 mL | 0.01 mL | 0.5 mL | Multi-pack |
£320.20
Pack of 500 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
12671470
|
Corning™
7078B-1 |
1 mL | 0.01 mL | 0.5 mL | Multi-pack |
£820.00
Pack of 500 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
11323845
|
Corning™
7077B-10 |
10 mL | 0.2 mL | 4 mL | Individually Wrapped |
£324.36
Pack of 200 |
In Stock
Log in to see stock. |
||
10292363
|
Corning™
7077B-50 |
50 mL | 0.5 mL | 3 mL | Individually Wrapped |
£237.91
Pack of 50 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
13043093
|
Corning™
7077B-50 |
50 mL | 0.5 mL | 3 mL | Individually Wrapped |
£118.96
Pack of 25 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||
10716323
|
Corning™
7077B-1 |
1 mL | 0.01 mL | 0.5 mL | Individually Wrapped |
£305.09
Pack of 500 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
13013093
|
Corning™
7077B-1 |
1 mL | 0.01 mL | 0.5 mL | Individually Wrapped |
£152.55
Pack of 250 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10130023
|
Corning™
7077B-5 |
5 mL | 0.1 mL | 3 mL | Individually Wrapped |
£559.02
Pack of 400 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10716693
|
Corning™
7077B-25 |
25 mL | 0.2 mL | 5 mL | Individually Wrapped |
£863.22
Pack of 400 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
13033093
|
Corning™
7077B-25 |
25 mL | 0.2 mL | 5 mL | Individually Wrapped |
£215.81
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||
10202613
|
Corning™
7077B-10 |
10 mL | 0.2 mL | 4 mL | Individually Wrapped |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
These shorty pipets, calibrated 'To Deliver' with blow-out, are ideal for use in confined areas. The 5, 10, and 25 mL sizes have tooled tops. Negative graduations on each pipet make it applicable for both serological and measuring procedures. Packaged in plastic bags so several can be removed without contaminating the entire pack. ISO color-coding is printed directly on the tear strip.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Cat. No. 7078B-50 features negative and reverse graduations.
Compliance
Calibrated To Deliver (TD) with blow-out.
Specifications
Yes | |
Borosilicate Glass | |
Round | |
Single-use |
Yes | |
Yes | |
Serological Pipet |