missing translation for 'onlineSavingsMsg'
Learn More
Pyridine, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10637684

Pyridine, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10637684
2.5 L, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10637684

Brand: Thermo Scientific Acros 290860025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717Show MoreShow Less
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Flash Point 17°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5085 to 1.5105
Quantity 2.5 L
Beilstein 20,181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15,8081
Specific Gravity 0.978
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Viscosity 0.95 mPa.s (20°C)
Formula Weight 79.1
Percent Purity 99+%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Pyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450989 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number290860025,290860010,290861000
Lot NumberA0460267 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number290860025,290860010,290861000
Lot NumberA0445552 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number290860025,290860100,290860010,290861000
Lot NumberA0403467 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number290860025,290860100,290860010,290861000
Lot NumberA0442652 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog Number290860025,290860100,290860010,290861000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.