missing translation for 'onlineSavingsMsg'
Learn More
Pyridine, ACS, 99.0+%, Thermo Scientific Chemicals Product Code.: 11490490

Pyridine, ACS, 99.0+%, Thermo Scientific Chemicals

Product Code. 11490490
500 mL
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11490490

Brand: Thermo Scientific Alfa Aesar 019378.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 0.002%, H2O 0.1%, Cl 0.001%, SO4 0.001%, NH3 0.002%, Cu 5ppm, Reducing substances P.T., Solubility in water P.T.

As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane (C5H5NBH3) and pyridine-sulfur trioxide (C5H5NSO3) are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717Show MoreShow Less
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point °C to 42°C
Color Colorless
Appearance Colorless liquid
Density 0.978
Boiling Point 115°C
Quantity 500 mL
Flash Point 20°C (68°F)
Ignition Point 482°C (900°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.51
Assay Percent Range 99.0% min.
Beilstein 103233
Solubility Information Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight 79.1
Grade ACS
Chemical Name or Material Pyridine, For ACS analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU01F048 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number019378.K2,019378.K7,019378.AP,019378.M1
Lot NumberM30L720 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number019378.K2,019378.K7,019378.AP,019378.M1
Lot NumberU01F050 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number019378.K2,019378.K7,019378.AP,019378.M1
Lot NumberJ15Y859 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number019378.K2,019378.K7,019378.AP,019378.M1
Lot NumberW30K925 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number019378.K2,019378.K7,019378.AP,019378.M1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine, ACS, 99.0+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.