Learn More
Thermo Scientific™ Quantech™ Fluorometer Narrow-Band Excitation and Emission Filters
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment.
£262.14 - £897.60
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11721111
|
Thermo Scientific™
LE1095X1 |
254 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11731111
|
Thermo Scientific™
LE1095X10 |
440 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11741111
|
Thermo Scientific™
LE1095X11 |
450 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11751111
|
Thermo Scientific™
LE1095X12 |
460 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11651359
|
Thermo Scientific™
LE1095X13 |
490 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11761111
|
Thermo Scientific™
LE1095X14 |
520 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11771111
|
Thermo Scientific™
LE1095X15 |
540 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781111
|
Thermo Scientific™
LE1095X16 |
590 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11791111
|
Thermo Scientific™
LE1095X17 |
680 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11701121
|
Thermo Scientific™
LE1095X3 |
297 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11711121
|
Thermo Scientific™
LE1095X30 |
360 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10324110
|
Thermo Scientific™
LE1095X35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||||
10739978
|
Thermo Scientific™
LE1095X4 |
313 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11771121
|
Thermo Scientific™
LE1095X6 |
365 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781121
|
Thermo Scientific™
LE1095X7 |
390 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11791121
|
Thermo Scientific™
LE1095X8 |
405 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11701131
|
Thermo Scientific™
LE1095X9 |
420 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Beschreibung
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment. Narrow band filters allow a narrow range of light to pass through the filter -- delivering the selectivity you need for accurate fluorescence assays.
Narrow band filters can be used to excite fluorescence or to measure a very small window of fluorescence emission. To select the proper filter for your assay, use the following guidelines:- There should be no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter
- A portion of the fluorescence excitation (absorption) spectrum near the peak falls in the wavelength range transmitted by the Narrow Band excitation filter
Spezifikation
Narrow Band Filter | |
Quantech |