missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

Kits enable quick and sensitive detection of low and high abundance RNA, and can distinguish between RNA and DNA, protein, and free nucleotides.

£71.50 - £334.00

Specifications

Detection Method Fluorescence
View More Specs

Products 8
Product Code Brand Quantity Assay Price Quantity & Availability  
Product Code Brand Quantity Assay Price Quantity & Availability  
10174653
View Documents
Invitrogen™
Q10210
100 assays RNA Quantification, BR
£102.00
Each
Please to purchase this item. Need a web account? Register with us today!
10266793
View Documents
Invitrogen™
Q10211
500 assays RNA Quantification, BR
£284.00
Each
Please to purchase this item. Need a web account? Register with us today!
15840210
View Documents
Invitrogen™
Q33223
100 assays RNA Quantification, ext range N/A N/A N/A
15810215
View Documents
Invitrogen™
Q33223
100 assays RNA Quantification, ext range
£110.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
15898627
View Documents
Invitrogen™
Q33224
500 assays RNA Quantification, ext range
£334.00
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
15859854
View Documents
Invitrogen™
Q33224
500 assays RNA Quantification, ext range N/A N/A N/A
10034622
View Documents
Invitrogen™
Q32852
100 assays RNA Quantification, HS
£96.25
1 set
Please to purchase this item. Need a web account? Register with us today!
10320093
View Documents
Invitrogen™
Q32855
500 assays RNA Quantification, HS
£284.00
500 assays
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Fluorescence
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.