missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly
Complete kit including all glass components necessary to perform a solid-liquid extraction. Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly includes an extractor, condenser, and an evaporating flask.
Brand: Quickfit™ 600RDSX
771.82 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Each assembly comprises the basic components for solid-liquid extraction : Flask/Soxhlet extractor/Condenser/Associated lids and clips
- Choice of assemblies with different extractor capacities

Specifications
Soxhlet Extraction Assembly | |
Pyrex Borosilicate Glass | |
Ideal for solid-liquid extraction |
600 mL | |
FR2L/5S Flask, EX5/75, DA5/75, JC75F Soxhlet extractor, CX5/25/SC condenser, and associated lids and clips |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly > Capacity: 600mL
Spot an opportunity for improvement?Share a Content Correction