Learn More
Quickfit™ Sloping Splash Head Adapter
Used in distillation assemblies.
Brand:  Quickfit SH7/43
447.35 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15507980
  • £621.00 / Pack of 5
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
Distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser
24/29 (Condenser), 24/29 (flask)
Sloping Splash Head
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Sloping Splash Head Adapter > Condenser Size: 24/29; Flask size: 24/29