Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
£77.30 - £202.00
Products 9
Product Code Brand Cone Size Standard Taper Joint No. Price Quantity & Availability  
Product Code Brand Cone Size Standard Taper Joint No. Price Quantity & Availability  
12028371
View Documents Promotion Details
Quickfit
MF17/1/SC
14/23 14/23
£77.30
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10440485
View Documents Promotion Details
Quickfit
MF18/2/SC
19/26 19/26
£94.25
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
12048371
View Documents Promotion Details
Quickfit
MF18/3/SC
24/29 19/26
£94.25
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
12058371
View Documents Promotion Details
Quickfit
MF18/33/SC
24/29 24/29
£97.20
Each
In Stock
to see stock.
Edge
Add to basket
12455316
View Documents Promotion Details
Quickfit
MF18/44/SC
29/32 29/32
£92.20
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
12465316
View Documents Promotion Details
Quickfit
MF18/5/SC
34/35 19/26
£97.95
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15507940
View Documents
Quickfit
MF18/4/SC
29/32 19/26
£188.00
Pack of 5
Estimated Shipment: 29-08-2025
to see stock.
Añadir a la cesta
15587930
Ver documentos
Quickfit
MF17/2/SC
19/26 14/23
£153.00
Pack of 5
Estimated Shipment: 29-07-2025
to see stock.
Añadir a la cesta
15597930
Ver documentos Detalles de la promoción
Quickfit
MF17/3/SC
NS 24/29 NS 24/29
£202.00
Pack of 5
Estimated Shipment: 13-06-2025
to see stock.
Añadir a la cesta
Descripción
Descripción
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
  • With sidearm for the attachment of rubber hose etc
  • Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Videos
SDS
Documentos
Documentos
Product Certifications
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Quickfit™ Tubing Adapter with Side-arm
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado