Learn More
Quickfit™ Screwthread Vacuum Receiver Adapter
Allows four flasks to be connected to a condenser outlet.
Brand: Quickfit RA11/SC
666.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Useful for collecting fractions when distilling under vacuum
- Each flask can be progressively filled by rotating the adapter
- Avoids problems of disturbing vacuum conditions when changing flasks
Specifications
Useful for collecting distillate fractions | |
Screw thread connector | |
Vacuum receiver adapter |
Flask | |
Borosilicate Glass |