Learn More
(+)-Quinidine, Thermo Scientific Chemicals
£18.90 - £113.00
Chemical Identifiers
CAS | 56-54-2 |
---|---|
Molecular Formula | C20H24N2O2 |
Molecular Weight (g/mol) | 324.42 |
MDL Number | MFCD00135581,MFCD00198096 |
InChI Key | LOUPRKONTZGTKE-LHHVKLHASA-N |
Synonym | quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine |
PubChem CID | 441074 |
ChEBI | CHEBI:28593 |
IUPAC Name | (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol |
SMILES | COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474254
|
Thermo Scientific Alfa Aesar
A12559.03 |
1 g |
£18.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484254
|
Thermo Scientific Alfa Aesar
A12559.06 |
5 g |
£31.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494254
|
Thermo Scientific Alfa Aesar
A12559.14 |
25 g |
£113.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
56-54-2 | |
324.42 | |
LOUPRKONTZGTKE-LHHVKLHASA-N | |
441074 | |
(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol |
C20H24N2O2 | |
MFCD00135581,MFCD00198096 | |
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine | |
CHEBI:28593 | |
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1 |
Specifications
56-54-2 | |
C20H24N2O2 | |
UN2811 | |
Light sensitive | |
quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine | |
LOUPRKONTZGTKE-LHHVKLHASA-N | |
(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol | |
441074 | |
324.42 |
171°C to 173°C | |
MFCD00135581,MFCD00198096 | |
91866 | |
14,8060 | |
Soluble in methanol,chloroform,alcohol (1g/36ml),benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water. | |
COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1 | |
324.42 | |
CHEBI:28593 | |
(+)-Quinidine |
Safety and Handling
GHS H Statement
H301
P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
H301-H317
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-279-0
missing translation for 'rtecsNumber' : VA4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only