Learn More
Quinuclidine, 97+%, Thermo Scientific Chemicals
£48.70 - £364.00
Chemical Identifiers
CAS | 100-76-5 |
---|---|
Molecular Formula | C7H13N |
Molecular Weight (g/mol) | 111.188 |
MDL Number | MFCD00006690 |
InChI Key | SBYHFKPVCBCYGV-UHFFFAOYSA-N |
Synonym | quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi |
PubChem CID | 7527 |
ChEBI | CHEBI:38420 |
IUPAC Name | 1-azabicyclo[2.2.2]octane |
SMILES | C1CN2CCC1CC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420788
|
Thermo Scientific Alfa Aesar
H54498.MD |
250 mg |
£48.70
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400788
|
Thermo Scientific Alfa Aesar
H54498.03 |
1 g |
£125.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410788
|
Thermo Scientific Alfa Aesar
H54498.06 |
5 g |
£364.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsQuinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.
Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Chemical Identifiers
100-76-5 | |
111.188 | |
SBYHFKPVCBCYGV-UHFFFAOYSA-N | |
7527 | |
1-azabicyclo[2.2.2]octane |
C7H13N | |
MFCD00006690 | |
quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi | |
CHEBI:38420 | |
C1CN2CCC1CC2 |
Specifications
100-76-5 | |
C7H13N | |
UN2811 | |
Air Sensitive | |
quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi | |
SBYHFKPVCBCYGV-UHFFFAOYSA-N | |
1-azabicyclo[2.2.2]octane | |
7527 | |
111.19 | |
Quinuclidine |
157°C to 160°C | |
MFCD00006690 | |
103111 | |
14,8081 | |
Soluble in alcohol,diethyl ether,water and organic solvents. | |
C1CN2CCC1CC2 | |
111.188 | |
CHEBI:38420 | |
≥97% |
Safety and Handling
GHS H Statement
H301-H310-H318-H315
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Causes skin irritation.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H310-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 202-887-1
missing translation for 'rtecsNumber' : CL5594625
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only