missing translation for 'onlineSavingsMsg'
Learn More
(R)-(+)-1-Boc-2-cyanopyrrolidine, 97%, Thermo Scientific™ Product Code.: 15464168

(R)-(+)-1-Boc-2-cyanopyrrolidine, 97%, Thermo Scientific™

Product Code. 15464168
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15464168

Brand: Thermo Scientific Alfa Aesar H57745.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 228244-20-0
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD06411382
InChI Key MDMSZBHMBCNYNO-MRVPVSSYSA-N
Synonym r-1-boc-2-cyanopyrrolidine, r-1-n-boc-2-cyano-pyrrolidine, r-tert-butyl 2-cyanopyrrolidine-1-carboxylate, tert-butyl 2r-2-cyanopyrrolidine-1-carboxylate, r-+-1-boc-2-cyanopyrrolidine, r-1-n-boc-2-cyanopyrrolidine, r-n-boc-2-pyrrolidinonitrile, r-+-1-boc-pyrrolidine-2-carbonitrile, r-2-cyano-pyrrolidine-1-carboxylic acid tert-butyl ester, pubchem18389Show MoreShow Less
PubChem CID 2756788
IUPAC Name tert-butyl (2R)-2-cyanopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C#N

Specifications

Melting Point 32°C to 37°C
Quantity 1 g
Sensitivity Light and moisture sensitive
Optical Rotation +105° (c=1 in Chloroform)
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material (R)-(+)-1-Boc-2-cyanopyrrolidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK06Y012 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(+)-1-Boc-2-cyanopyrrolidine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.