missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals Product Code.: 15489525

(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals

Product Code. 15489525
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15489525

Brand: Thermo Scientific Alfa Aesar H52037.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517905-93-0
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD03428045
InChI Key DRESTDPDCGPKNI-UHFFFAOYNA-N
Synonym r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-r-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24026, fmoc-beta-phe 2-no2-oh, fmoc-d-?-phe 2-no2-oh, n-fmoc-2-nitro-l-beta-phenylalanine, fmoc-r-3-amino-3-2-nitro-phenyl-propionic acid, r-3-fmoc-amino-3-2-nitrophenyl propionic acidShow MoreShow Less
PubChem CID 2761743
IUPAC Name 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
SMILES OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 170°C to 172°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Optical Rotation −182° (c=1 in DMF)
Formula Weight 432.43
Percent Purity 98%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS13K019 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog NumberH52037.06,H52037.03
Lot NumberZ19J056 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberH52037.MD,H52037.06,H52037.03
Lot NumberZ04F034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52037.MD,H52037.06,H52037.03
Lot NumberK16W024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52037.MD,H52037.06,H52037.03
Lot NumberR30D031 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52037.MD,H52037.06,H52037.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.