Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Brand: Thermo Scientific™ 4000310
This item is not returnable.
View return policy
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details
Specifications
Silver | |
All CryoMed controlled-rate freezers | |
Skin Rack |
48 Skin Packets (2 level) | |
48 Skin Packets (2 level) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Cryopreservation Storage Equipment Brochure
File size: 4.4 MB
CryoMed Controlled-Rate Freezers
Version: FEB.2016
File size: 3.6 MB
Product Content Correction
Product Title
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer > Skin rack, 2 level (holds 48 skin packets)
Spot an opportunity for improvement?Share a Content Correction