Learn More
Thermo Scientific™ MarqMetrix Replacement FlowCell BallProbe
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
Brand: Thermo Scientific™ 810-02861-01
This item is not returnable.
View return policy
Description
- Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
Specifications
MarqMetrix All-In-One Process Raman Analyzer | |
Replacement FlowCell BallProbe |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Thermo Scientific™ MarqMetrix Replacement FlowCell BallProbe > Replacement Part
Spot an opportunity for improvement?Share a Content Correction