Learn More
GIBCO™ Rat (Sprague Dawley) Cytosol
Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats. Total protein concentration: 20mg/mL
Brand:  GIBCO™ RTCYPL
Product Code. 10382980
  • £42.20 / 1mL
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Specifications
Pooled
Frozen
1 mL
Rat
Rat Cytosol
Tube
Dry Ice
Male
Liver
Research Grade
20 mg/mL
Store between -60°C and -85°C.
Cytosol
Gibco™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberRT062-C Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberRTCYPL
Lot NumberRT062-D Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberRTCYPL
Lot NumberRT062-E Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberRTCYPL
Lot NumberRT062-F Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberRTCYPL
Lot NumberRT062-A Certificate TypeCertificate of Analysis Date11/02/2020 Catalog NumberRTCYPL
Special Offers
Special Offers
Product Content Correction
Product Title
GIBCO™ Rat (Sprague Dawley) Cytosol > Rat (Sprague Dawley) Cytosol