Learn More
RCAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12255-1-AP-150UL
 View more versions of this product
Product Code. 16809863
  • £349.00 / 150µL
Explore more special offers
Description
Description
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5′-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.
Specifications
Specifications
RCAS1
Polyclonal
Unconjugated
EBAG9
EB9, EBAG9, PDAF, RCAS1
Rabbit
Antigen Affinity Chromatography
RUO
299864, 55960, 9166
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00559, Q5PQP2, Q9D0V7
Ebag9
RCAS1 Fusion Protein Ag2905
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RCAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated