missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S2, was expressed in the hosts and tags indicated.
£281.57 - £2410.48
Specifications
Accession Number | NP_414711.1 |
---|---|
Gene ID (Entrez) | 947874 |
Name | 30S ribosomal protein S2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065581
|
enQuireBio™
QP8615-ec-10ug |
10 μg | |||||||
16005591
|
enQuireBio™
QP8615-ec-50ug |
50 μg | |||||||
16055581
|
enQuireBio™
QP8615-ec-100ug |
100 μg | |||||||
16085581
|
enQuireBio™
QP8615-ec-200ug |
200 μg | |||||||
16095581
|
enQuireBio™
QP8615-ec-500ug |
500 μg | |||||||
16075581
|
enQuireBio™
QP8615-ec-1mg |
1 mg | |||||||
Specifications
NP_414711.1 | |
30S ribosomal protein S2 Protein | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |
947874 | |
Research Use Only | |
rpsB | |
Recombinant Protein | |
E. coli | |
ATVSMRDMLK AGVHFGHQTR YWNPKMKPFI FGARNKVHII NLEKTVPMFN EALAELNKIA SRKGKILFVG TKRAASEAVK DAALSCDQFF VNHRWLGGML TNWKTVRQSI KRLKDLETQS QDGTFDKLTK KEALMRTREL EKLENSLGGI KDMGGLPDAL FVIDADHEHI AIKEANNLGI PVFAIVDTNS DPDGVDFVIP GNDDAIRAVT LYLGAVAATV REGRSQDLAS QAEESFVEAE | |
Greater than 90% as determined by SDS-PAGE. |