missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) cytomegalovirus (HCMV) Glycoprotein B / gB, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | YP_401713.1 |
---|---|
Gene ID (Entrez) | 3783680 |
Name | cytomegalovirus (HCMV) Glycoprotein B / gB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077950
|
enQuireBio™
QP7275-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097950
|
enQuireBio™
QP7275-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087950
|
enQuireBio™
QP7275-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007960
|
enQuireBio™
QP7275-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017960
|
enQuireBio™
QP7275-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027960
|
enQuireBio™
QP7275-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
YP_401713.1 | |
cytomegalovirus (HCMV) Glycoprotein B / gB Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783680 | |
Research Use Only | |
gB | |
Recombinant Protein | |
E. coli | |
QTPEQPAPPA TTVQPTATRQ QTSFPFRVCE LSSHGDLFRF SSDIQCPSFG TRENHTEGLL MVFKDNIIPY SFKVRSYTKI VTNILIYNGW YADSVTNRHE EKFSVDSYET DQMDTIYQCY NAVKMTKDGL TRVYVDRDGV NITVNLKPTG GLANGVRRYA SQTELYDAPG WLIWTYRTRT TVNCLITDMM AKSNSPFDFF VTTTGQTVEM SPFYDGKNKE TFHERADSFH VRTNYKIV | |
Greater than 90% as determined by SDS-PAGE. |